Anti-RASSF1 Mouse monoclonal antibody [clone: 8E4]

Lieferant: Abcam
ABCAAB119423-100EA 781 EUR
ABCAAB119423-100
Anti-RASSF1 Mouse monoclonal antibody [clone: 8E4]
Type: Primary
Antigen: RASSF1 (Ras association (RalGDS/AF-6) domain family member 1)
Clonality: monoclonal
Clone: 80000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR