Anti-CBP Tag Rabbit Polyclonal Antibody

Lieferant: Abcam
AB119488-100
ABCAAB119488-100EA 848 EUR
ABCAAB119488-100
Anti-CBP Tag Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CBP Tag
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR