Anti-OPA1 Mouse Monoclonal Antibody [clone: 1E81D9]

Lieferant: Abcam
AB119685-100
ABCAAB119685-100EA 891 EUR
ABCAAB119685-100
Anti-OPA1 Mouse Monoclonal Antibody [clone: 1E81D9]
Antikörper
Type: Primary
Antigen: OPA1 (optic atrophy 1 (autosomal dominant))
Clonality: monoclonal
Clone: 1E81D9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR