Anti-ACTR2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB119766-100EA 840 EUR
ABCAAB119766-100
Anti-ACTR2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ACTR2 (ARP2 actin-related protein 2 homolog (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR