Anti-HES1 Mouse monoclonal antibody [clone: 2D2]

Lieferant: Abcam
ABCAAB119776-100EA 739 EUR
ABCAAB119776-100
Anti-HES1 Mouse monoclonal antibody [clone: 2D2]
Type: Primary
Antigen: HES1 (hes family bHLH transcription factor 1)
Clonality: monoclonal
Clone: 2D2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR