Anti-CD34 Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: 581]

Lieferant: Abcam
ABCAAB119840-50EA 834 EUR
ABCAAB119840-50
Anti-CD34 Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: 581]
Antikörper
Type: Primary
Antigen: CD34 (CD34 molecule)
Clonality: monoclonal
Clone: 581
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR