Anti-RAB18 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB119900-100
ABCAAB119900-100EA 896 EUR
ABCAAB119900-100
Anti-RAB18 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RAB18 (RAB18, member RAS oncogene family)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR