Anti-MUC19 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB121014-200EA 535 EUR
ABCAAB121014-200
Anti-MUC19 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MUC19 (mucin 19, oligomeric)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR