Anti-THSD7A Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB121122-100EA 839 EUR
ABCAAB121122-100
Anti-THSD7A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: THSD7A (thrombospondin, type I, domain containing 7A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human