Anti-FAM181A Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB121144-100EA 816 EUR
ABCAAB121144-100
Anti-FAM181A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FAM181A (family with sequence similarity 181, member A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human