Anti-LAMTOR1 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB121157-100EA 816 EUR
ABCAAB121157-100
Anti-LAMTOR1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LAMTOR1 (late endosomal/lysosomal adaptor, MAPK and MTOR activator 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human