Anti-LRCH2 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB121182-100EA 816 EUR
ABCAAB121182-100
Anti-LRCH2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LRCH2 (leucine-rich repeats and calponin homology (CH) domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human