Anti-FCGBP Rabbit polyclonal antibody unconjugated
Lieferant: Abcam
ABCAAB121199-100EA 835 EUR
ABCAAB121199-100
Anti-FCGBP Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: FCGBP (Fc fragment of IgG binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human