Anti-PLEKHA2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB121218-100EA 816 EUR
ABCAAB121218-100
Anti-PLEKHA2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PLEKHA2 (pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR