Anti-CT83 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB121219-100EA 834 EUR
ABCAAB121219-100
Anti-CT83 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: CT83 (cancer/testis antigen 83)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR