Anti-EXD2 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB121236-100EA 816 EUR
ABCAAB121236-100
Anti-EXD2 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: EXD2 (exonuclease 3'-5' domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR