Anti-MCPH1 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB121277-100
ABCAAB121277-100EA 902 EUR
ABCAAB121277-100
Anti-MCPH1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MCPH1 (microcephalin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR