Anti-SHROOM4 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB121316-100EA 816 EUR
ABCAAB121316-100
Anti-SHROOM4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SHROOM4 (shroom family member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR