Anti-TMIGD2 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB121333-100
ABCAAB121333-100EA 881 EUR
ABCAAB121333-100
Anti-TMIGD2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TMIGD2 (transmembrane and immunoglobulin domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR