Anti-MRPL52 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB121366-100EA 816 EUR
ABCAAB121366-100
Anti-MRPL52 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MRPL52 (mitochondrial ribosomal protein L52)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR