Anti-ZDHHC6 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB121423-100EA 816 EUR
ABCAAB121423-100
Anti-ZDHHC6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZDHHC6 (zinc finger, DHHC-type containing 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human