Anti-TMEM223 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB121434-100EA 816 EUR
ABCAAB121434-100
Anti-TMEM223 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TMEM223 (transmembrane protein 223)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human