Anti-C19orf59 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB121447-100EA 816 EUR
ABCAAB121447-100
Anti-C19orf59 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: C19orf59 (chromosome 19 open reading frame 59)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human