Anti-LRIG2 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB121472-100
ABCAAB121472-100EA 877 EUR
ABCAAB121472-100
Anti-LRIG2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LRIG2 (leucine-rich repeats and immunoglobulin-like domains 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR