Anti-TMEM45B Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB121488-100EA 816 EUR
ABCAAB121488-100
Anti-TMEM45B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TMEM45B (transmembrane protein 45B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human