Anti-SLFN5 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB121537-100EA 835 EUR
ABCAAB121537-100
Anti-SLFN5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SLFN5 (schlafen family member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human