Anti-ARHGAP23 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB121617-100EA 816 EUR
ABCAAB121617-100
Anti-ARHGAP23 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ARHGAP23 (Rho GTPase activating protein 23)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR