Anti-TANC2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB121745-100EA 816 EUR
ABCAAB121745-100
Anti-TANC2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TANC2 (tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR