Anti-ZNF653 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB121769-100
ABCAAB121769-100EA 857 EUR
ABCAAB121769-100
Anti-ZNF653 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZNF653 (zinc finger protein 653)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR