Anti-ALOX12B Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB121785-100EA 816 EUR
ABCAAB121785-100
Anti-ALOX12B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ALOX12B (arachidonate 12-lipoxygenase, 12R type)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR