Anti-KIF21B Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB121931-100EA 816 EUR
ABCAAB121931-100
Anti-KIF21B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KIF21B (kinesin family member 21B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human