Anti-SRGAP2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB121977-100EA 834 EUR
ABCAAB121977-100
Anti-SRGAP2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SRGAP2 (SLIT-ROBO Rho GTPase activating protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR