Anti-SPAG17 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB122008-100EA 816 EUR
ABCAAB122008-100
Anti-SPAG17 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SPAG17 (sperm associated antigen 17)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR