Anti-AHDC1 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB122009-100
ABCAAB122009-100EA 857 EUR
ABCAAB122009-100
Anti-AHDC1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AHDC1 (AT hook, DNA binding motif, containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR