Anti-LRP5L Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB122013-100EA 816 EUR
ABCAAB122013-100
Anti-LRP5L Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LRP5L (low density lipoprotein receptor-related protein 5-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human