Anti-ZGRF1 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB122126-100EA 816 EUR
ABCAAB122126-100
Anti-ZGRF1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZGRF1 (zinc finger, GRF-type containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human