Anti-EPG5 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB122186-100EA 816 EUR
ABCAAB122186-100
Anti-EPG5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EPG5 (ectopic P-granules autophagy protein 5 homolog (C. elegans))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human