Anti-MMGT1 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB122202-100
ABCAAB122202-100EA 877 EUR
ABCAAB122202-100
Anti-MMGT1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MMGT1 (membrane magnesium transporter 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR