Anti-ECM2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB122268-100EA 816 EUR
ABCAAB122268-100
Anti-ECM2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ECM2 (extracellular matrix protein 2, female organ and adipocyte specific)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR