Anti-CCDC39 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB122269-100EA 816 EUR
ABCAAB122269-100
Anti-CCDC39 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CCDC39 (coiled-coil domain containing 39)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR