Anti-ZSWIM6 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB122301-100EA 816 EUR
ABCAAB122301-100
Anti-ZSWIM6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZSWIM6 (zinc finger, SWIM-type containing 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human