Anti-ANKRD23 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB122320-100
ABCAAB122320-100EA 877 EUR
ABCAAB122320-100
Anti-ANKRD23 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ANKRD23 (ankyrin repeat domain 23)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR