Anti-DNAH6 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB122333-100
ABCAAB122333-100EA 857 EUR
ABCAAB122333-100
Anti-DNAH6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DNAH6 (dynein, axonemal, heavy chain 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR