Anti-STT3B Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB122351-100EA 816 EUR
ABCAAB122351-100
Anti-STT3B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: STT3B (STT3B, subunit of the oligosaccharyltransferase complex (catalytic))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human