Anti-TTC7A Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB122363-100EA 816 EUR
ABCAAB122363-100
Anti-TTC7A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TTC7A (tetratricopeptide repeat domain 7A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR