Anti-BOD1L1 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB122380-100EA 835 EUR
ABCAAB122380-100
Anti-BOD1L1 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: BOD1L1 (biorientation of chromosomes in cell division 1-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR