Anti-ATRNL1 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB122437-100EA 816 EUR
ABCAAB122437-100
Anti-ATRNL1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ATRNL1 (attractin-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human