Anti-EPYC Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB122449-100EA 835 EUR
ABCAAB122449-100
Anti-EPYC Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EPYC (epiphycan)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human