Anti-SVIP Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB122590-100EA 816 EUR
ABCAAB122590-100
Anti-SVIP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SVIP (small VCP/p97-interacting protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human