Anti-IGLON5 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB122763-100EA 837 EUR
ABCAAB122763-100
Anti-IGLON5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IGLON5 (IgLON family member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR